Search Thermo Fisher Scientific
HeLa cell transfected with pShooter pCMV/myc/mito/GFP, then fixed and permeabilized. Green-fluorescent protein (GFP) localized in the mitochondria was labeled with anti-GFP mouse IgG2a (Cat. no. A11120) and detected with orange-fluorescent Alexa Fluor® 555 goat anti–mouse IgG (CAt. no. A21422), which colocalized with the dim GFP fluorescence. F-actin was labeled with green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379), and the nucleus was stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490). The sample was mounted using ProLong® Gold antifade reagent (Cat. no. P36930). Some GFP fluorescence is retained in the mitochondria after fixation (top), but immunolabeling and detection greatly improve visualization (bottom).
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›