Providing 20 Years of RNA Essentials.
Ambion RNA essentials are designed to meet the needs of researchers working with RNA, offering key products like:
  • 核酸酶抑制剂和去除核酸酶污染试剂
  • 无核酸酶的缓冲液和塑料制品
  • 样本收集和 RNA 稳定液
  • 梯度 RNA 分子量标准 (RNA ladder)
  • Northern 印迹试剂
RNA can be a difficult molecule to work with and is readily degraded by RNases that are found in a variety of environmental sources, including fingerprints. By minimizing nuclease contamination and employing the most stringent quality specifications, Ambion product essentials are designed to succeed in less-than-ideal conditions. This is referred to as "building robustness" into the products, which continues to be a primary focus of our product development today.
Ambion RNA essentials
Nuclease-Free Microfuge, Conical, PCR Tubes and Caps
Ambion Tubes & Caps are guaranteed RNase- and DNase-free.
Each lot of tubes is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free. The standard testing calls for incubation of labeled nucleic acid in randomly sampled tubes at 37°C for 14 to 16 hr. The nucleic acid from each tube is then analyzed for degradation by polyacrylamide gel electrophoresis
  • AM12225 - Thin-walled, frosted lid, RNase-free PCR tubes (1000 x 0.2ml)
  • AM12230 - PCR tubes & caps, RNase-free, 0.2 ml (125 strips x 8-tubes)
  • AM12250 - Thin-walled, domed cap, RNase-free PCR tubes (1000 x 0.5 ml)
  • AM12275 - Thin-walled, frosted lid, RNase-free PCR tubes (1000 x 0.5 ml)
  • AM12300 - RNase-free microfuge tubes (1000 x 0.5 ml)
  • AM12350 - Non-stick RNase-free microfuge tubes (500 x 0.5 ml)
  • AM12400 - RNase-free microfuge tubes (500 x 1.5 ml)
  • AM12425 - RNase-free microfuge tubes (500 x 2.0 ml)
  • AM12475 - Non-stick RNase-free microfuge tubes 250 x 2.0 ml)
  • AM12500 - Conical tubes (500 x 15 ml)
  • AM12501 - Conical tubes (200 x 50 ml)
Nuclease-Free Pipette Tips
Each lot of Ambion Tips is subjected to rigorous testing and is certified to be nuclease-free.
Ambion Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals.
  • AM12650 - RNase-free tips, 200 μl size (Ten 8 x 12 racks)
  • AM12660 - RNase-free tips, 1000 μl size (Ten racks x 100 tips each)
  • AM12635 - Barrier (filter) tips, 10 μl size - Eppendorf (Ten 8 x 12 racks)
  • AM12645 - Barrier (filter) tips, 20 μl size (Ten 8 x 12 racks)
  • AM12648 - Barrier (filter) tips, 100 μl size (Ten 8 x 12 racks)
  • AM12655 - Barrier (filter) tips, 200 μl size (Ten 8 x 12 racks)
  • AM12665 - Barrier (filter) tips, 1000 μl size (Ten racks x 100 tips each)
Surface Decontaminants
Ambion RNaseZap is a surface decontamination solution that removes RNase contamination from glass and plastic surfaces. It’s ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free.
Ambion ElectroZap™ is specially formulated to instantaneously eliminate RNase contamination from electrodes.
  • AM9780 - RNaseZap, 250 ml
  • AM9780M - RNaseZap with Manual, 250 ml
  • AM9782 - RNaseZap, 6 x 250 ml
  • AM9782M - RNaseZap with manual, 6 x 250 ml
  • AM9784 - RNaseZap, 4 L
  • AM9786 - RNaseZap Wipes, 100 sheets
  • AM9786M - RNaseZap Wipes with Manual, 100 sheets
  • AM9788 - RNaseZap Wipes Refill, 300 sheets
  • AM9788M - RNaseZap Wipes Refill with manual, 300 sheets
  • AM9785 - ElectroZap™, 250 ml
Nuclease Inhibitors
SUPERase•In™ RNase Inhibitor is a protein based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases including RNase A, B, C, 1 and T1.
Ambion ANTI-RNase is a protein ribonuclease inhibitor that noncovalently binds and inactivates neutral pancreatic RNase A–type enzymes.
Ambion RNAsecure™ (patents pending) is a unique non-enzymatic reagent that will irreversibly inactivate RNases in solution.
  • AM2694 - SUPERase•In™ (20 U/µl) (2500 U)
  • AM2696 - SUPERase•In™ (20 U/µl) (10,000 U)
  • AM2690 - ANTI-RNase (15-30 U/µl) (2500 U)
  • AM2692 - ANTI-RNase (15-30 U/µl) (10,000 U)
  • AM7006 - RNAsecure™ Reagent (10 ml)
无核酸酶水
Ambion Nuclease-free water has not been treated with DEPC; residual DEPC may negatively impact some applications (i.e. oocyte injection). It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
  • 4387936 - Nuclease-free Water (not DEPC-treated), 4 L
  • AM9932 - Nuclease-free Water (not DEPC-treated), 1 L
  • AM9937 - Nuclease-free Water (not DEPC-treated), 10 x 50 ml
  • AM9939 - Nuclease-free Water (not DEPC-treated), 5 x 100 ml
  • AM9930 - Nuclease-free Water (not DEPC-treated), 500 ml
  • AM9938 - Nuclease-free Water (not DEPC-treated), 100 ml
  • AM9916 - DEPC-treated Water, 5 x 100 ml
缓冲液和化学品
Gel running solutions, RNA storage solutions, buffer kit, coprecipitants for nucleic acid precipitation, and a wide selection of individual buffers.
  • AM9855G - 1 M Tris, pH 8.0, 100 ml
  • AM9856 - 1 M Tris, pH 8.0, 500 ml
  • AM9849 - TE, pH 8.0, 500 ml
  • AM9858 - TE, pH 8.0, 1 L
  • AM9624 - PBS 10X, pH 7.4, 500 ml
  • AM9625 - PBS 10X, pH 7.4, 1 L
  • AM9680 - Salmon Sperm DNA (sheared, 10 mg/ml), 10 x 10 mg
  • AM9720 - Acid-Phenol:Chloroform, pH 4.5 (with IAA, 125:24:1), 100 ml
  • AM9722 - Acid-Phenol:Chloroform, pH 4.5 (with IAA, 125:24:1), 400 ml
  • AM9510 - Glycogen (5 mg/ml), 5 x 1 ml
  • AM9515 - GlycoBlue™ (15 mg/ml) 300 µl
  • AM9516 - GlycoBlue™ (15 mg/ml) 5 x 0.3 ml
  • AM2546 - Proteinase K Solution (20 mg/ml), 1 ml
  • AM2548 - Proteinase K Solution (20 mg/ml), 5 x 1.25 ml
Sample Collection Stabilization
RNA Later is an aqueous, nontoxic tissue storage reagent that rapidly permeates tissues to stabilize and protect cellular RNA.
  • Single reagent immediately inactivates RNases and stabilizes RNA within tissues or cells
  • No need to freeze samples in liquid nitrogen or rush samples back to the lab freezer
  • Eliminates the need to freeze and grind most tissue samples
  • Perfect for tissue collection "in the field"
  • RNA is stable for 1 day at 37°C, 1 week at 25°C, 1 month at 4°C or indefinitely at -20°C
  • Compatible with many RNA isolation procedures including most RNA isolation kits
Ambion RNAlater-ICE is a novel reagent for transitioning frozen tissue to a state enabling easy extraction of high-quality RNA; frozen tissues are simply submerged in Ambion RNAlater-ICE and allowed to thaw overnight at –20°C. Once thawed, the tissues can be processed like fresh tissues using standard RNA isolation procedures. No more laborious grinding of frozen tissue to safeguard the RNA.
  • AM7020 - RNAlater, 100 ml
  • AM7021 - RNAlater, 500 ml
  • AM7024 - RNAlater, 250 ml
  • AM7023 - RNAlater, 20 bottles x 5 ml
  • AM7022 - RNAlater, 50 tubes x 1.5 ml
  • AM7030 - RNAlater-ICE, 25 ml
RNA Ladder
Ambion RNA Millennium™ Markers are designed to provide accurate size determination of single-stranded RNA transcripts from 0.5 to 9 kb and can be used in any northern protocol.
Ambion RNA Century™-Plus Markers are a set of 7 in vitro transcripts of 100, 200, 300, 400, 500, 750, and 1000 bases.
Ambion RNA Century™ Markers are a set of 5 in vitro transcripts of 100, 200, 300, 400, and 500 bases.
The Ambion Decade™ Marker System produces a set of radiolabeled RNA molecules of 150, 100, 90, 80, 70, 60, 50, 40, 30, 20, and 10 nucleotides. The molecular weight markers are ideally suited for experiments where RNA molecules less than 150 nucleotides in length are being studied.
  • AM7150 - RNA Millennium™ Markers (25 lanes)
  • AM7145 - RNA Century™-Plus Markers (50 µg)
  • AM7140 - RNA Century™ Markers (50 µg)
  • AM7778 - Decade™ Markers (10 rxns)
Northern Blotting Reagents
The Ambion NorthernMax-Gly Kit is an alternative to formaldehyde-based systems for northern analysis and uses a glyoxal/DMSO Loading Solution for sample denaturation.
The Ambion NorthernMax Kit contains a complete set of RNase-free reagents for running formaldehyde-based northern analysis. This kit is ideal for the investigator who wants to use familiar reagents while taking advantage of NorthernMax Kit's high sensitivity. Sufficient reagents are provided to process 1000 cm2 membrane.
  • AM1946 - NorthernMax-Gly Kit (1000 cm2)
  • AM1940 - NorthernMax Kit (1000 cm2)
  • AM8670 - ULTRAhyb Ultrasensitive Hybridization Buffer (125 ml)
仅供科研使用,不可用于诊断目的。