Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human BLMH quantitates human BLMH in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BLMH.
The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : BH, BLMH, BMH
Gene ID : (Human) 642
Gene symbol : BLMH
Protein Aliases : BH, Bleomycin hydrolase, BLM hydrolase
UniProt ID (Human) Q13867
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support