Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human FCRL3/FcRH3 quantitates human FCRL3/FcRH3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FCRL3/FcRH3.
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CD307c, FCRH3, FCRL3, IFGP3, IRTA3, SPAP2
Gene ID : (Human) 115352
Gene symbol : FCRL3
Protein Aliases : CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FcR-like protein 3, FcRH3, hIFGP3, IFGP family protein 3, Immune receptor translocation-associated protein 3, immunoglobulin superfamily receptor translocation associated protein 3, MAIA, SH2 domain-containing phosphatase anchor protein 2
UniProt ID (Human) Q96P31
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support