Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human IL-1 R6 quantitates human IL-1 R6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-1 R6.
The protein encoded by this gene is a member of the interleukin 1 receptor family. An experiment with transient gene expression demonstrated that this receptor was incapable of binding to interleukin 1 alpha and interleukin 1 beta with high affinity. This gene and four other interleukin 1 receptor family genes, including interleukin 1 receptor, type I (IL1R1), interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 1 (IL1RL1), and interleukin 18 receptor 1 (IL18R1), form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : IL-1Rrp2, IL-36R, IL1R-rp2, IL1RL2, IL1RRP2
Gene ID : (Human) 8808
Gene symbol : IL1RL2
Protein Aliases : IL-1 receptor-related protein 2, IL-1Rrp2, IL-36 receptor, IL-36R, interleukin 1 receptor-like 2, Interleukin-1 receptor-like 2, Interleukin-1 receptor-related protein 2
UniProt ID (Human) Q9HB29
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support