Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human MEPE/OF45 quantitates human MEPE/OF45 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MEPE/OF45.
Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family. MEPE promotes renal phosphate excretion and modulates mineralization. It can be phosphorylated by secreted protein kinases. MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines. It is a 525-amino acid protein with a calculated molecular weight of 58 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : MEPE, OF45
Gene ID : (Human) 56955
Gene symbol : MEPE
Protein Aliases : Matrix extracellular phosphoglycoprotein, matrix, extracellular phosphoglycoprotein with ASARM motif (bone), OF45, Osteoblast/osteocyte factor 45, Osteoregulin
UniProt ID (Human) Q9NQ76
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support