Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Human Serpin A3/Alpha-1-Antichymotrypsin quantitates human Serpin A3/Alpha-1-Antichymotrypsin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin A3/Alpha-1-Antichymotrypsin.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : AACT, ACT, GIG24, GIG25, SERPINA3
Gene ID : (Human) 12
Gene symbol : SERPINA3
Protein Aliases : ACT, Alpha-1-antichymotrypsin, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
UniProt ID (Human) P01011
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support