Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Human Soluble Tumor Necrosis Factor Receptor 1 (sTNFR1-60 kDa) ELISA quantitates sTNFR1 in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu sTNFR1.
Principle of the method
The Human sTNFR1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
TNFR1 (Tumor necrosis factor receptor 1) belongs to the tumor necrosis factor superfamily, and is one of the major TNF-alpha receptors. TNFR1 plays an important role in mediating, in cytokine mediated signaling, positive regulation of the NF-Kb pathway, positive regulation of angiogenesis, and negative regulation of gene expression. The extracellular domain of TNFR1 is also released into the circulatory system as soluble TNFR1 (sTNFR1). In humans, the TNFR1 gene is located on chromosome 12. Anti-apoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with TNFR1, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of TNFR1 were found to be associated with the autosomal dominant periodic fever syndrome, and the impaired receptor clearance is thought to be a mechanism of the disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CD120a, FPF, MS5, p55, p55-R, p60, TBP1, TNF-R, TNF-R-I, TNF-R55, TNFAR, TNFR1, TNFR1-d2, TNFR55, TNFR60, TNFRSF1A
Gene ID : (Human) 7132
Gene symbol : TNFRSF1A
Protein Aliases : CD120a, p55, p60, TNF-R1, TNF-RI, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, Tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
UniProt ID (Human) P19438
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support