Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Human TYRO-3 ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of human TYRO-3.
TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, signal transduction, cell adhesion. and cytoskeleton reorganization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : BYK, DTK, Etk-2, Rek, RSE, SKY, TIF, TYRO3
Gene ID : (Human) 7301
Gene symbol : TYRO3
Protein Aliases : TYRO3, BYK, Dtk, RSE, Sky, Tif, Etk-2, Rek, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase BYK, Tyrosine-protein kinase DTK, Tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY, Tyrosine-protein kinase TIF
UniProt ID (Human) Q06418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support