Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human Transglutaminase 2/TGM2 quantitates human Transglutaminase 2/TGM2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Transglutaminase 2/TGM2.
Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer's disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : TG(C), TGC, TGM2
Gene ID : (Human) 7052
Gene symbol : TGM2
Protein Aliases : C polypeptide, protein-glutamine-gamma-glutamyltransferase, Erythrocyte transglutaminase, G(h), Heart G alpha(h), hhG alpha(h), Isopeptidase TGM2, Protein G alpha(h), Protein-glutamine deamidase TGM2, Protein-glutamine dopaminyltransferase TGM2, Protein-glutamine gamma-glutamyltransferase 2, Protein-glutamine histaminyltransferase TGM2, Protein-glutamine noradrenalinyltransferase TGM2, Protein-glutamine serotonyltransferase TGM2, TG(C), TG2, TGase C, TGase H, TGase II, TGase-2, Tissue transglutaminase, Transglutaminase C, Transglutaminase H, Transglutaminase II, Transglutaminase-2, tTG
UniProt ID (Human) P21980
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support