Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mouse Gremlin-1 quantitates mouse Gremlin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Gremlin-1.
In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Cktsf1b1, Drm, Grem, Grem1, ld
Gene ID : (Mouse) 23892
Gene symbol : Grem1
Protein Aliases : Cysteine knot superfamily 1, BMP antagonist 1, Down-regulated in Mos-transformed cells protein, down-regulated in v-mos-transformed cells, Gremlin-1
UniProt ID (Mouse) O70326
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support