Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human STAT5A/B in cell lysates. Detection of mouse and rat STAT5A/B is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor-associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated by, and mediates the responses of, many cell ligands, such as IL-2, IL-3, IL-7, GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of this protein in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of this gene is found to induce the expression of BCL2L1/ BCL-X(L), which suggests the antiapoptotic function of this gene in cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : MGF, STAT5, STAT5A, STAT5B
Gene ID : (Human) 6776, (Human) 6777
Gene symbol : STAT5A, STAT5B
Protein Aliases : Signal transducer and activator of transcription 5A, Signal transducer and activator of transcription 5B, transcription factor STAT5B
UniProt ID (Human) P42229, (Human) P51692
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support