Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human and mouse p38 MAPK in cell lysates. Detection of rat p38 MAPK is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
MAPK14 (p38 alpha) is a member of the stress-activated protein kinase class of MAPKs. The p38 MAPK cascade regulates a variety of cellular responses to stress, inflammation, and other signals. p38 MAPK is relatively inactive in the nonphosphorylated form and becomes rapidly activated by dual phosphorylation of a Thr-Gly-Tyr motifs. There are four isoforms of p38 MAPK, which differ in their tissue expression and affinity for upstream activators and downstream effectors. When cells are exposed to tumor necrosis factor, interleukin-1, heat shock, or other activating stimuli, activation of MAPK kinase-3 occurs by phosphorylation. Activated MAPK kinase-3/6 phosphorylate each residue of Thr180 and Tyr182 in p38 MAPK. Phospho-p38 MAPK activates ATF-2, CHOP-1, MEF-2 and other transcription factors through phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Crk1, CSBP, CSBP1, CSBP2, CSPB1, EXIP, MAPK14, MXI2, p38, p38-alpha, p38a, p38ALPHA, p38MAPK, PRKM14, PRKM15, RK, SAPK2A
Gene ID : (Human) 1432, (Mouse) 26416
Gene symbol : MAPK14, Mapk14
Protein Aliases : CRK1, CSAID-binding protein, Csaids binding protein, cytokine suppressive anti-inflammatory drug binding protein, cytokine suppressive anti-inflammatory drug binding protein 1, Cytokine suppressive anti-inflammatory drug-binding protein, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, MAX-interacting protein 2, mitogen activated protein kinase 14, Mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, p38 alpha, p38 MAP kinase, p38 MAP kinase alpha, p38 MAPK, p38 mitogen activated protein kinase, p38alpha Exip, SAPK2a, Stress-activated protein kinase 2a, tRNA synthetase cofactor p38
UniProt ID (Human) Q16539, (Mouse) P47811
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support