Easy exome sequencing
Harness the power of the Ion AmpliSeq™ Exome solution to identify relevant variants in your research
Exome sequencing case study
Learn more about the Ion AmpliSeq™ Exome solution >
Exome sequencing overview
What is exome sequencing?
Exome sequencing is a targeted sequencing approach that is restricted to the protein-coding regions of genomes. The exome is estimated to encompass approximately 1% of the genome, yet contains approximately 85% of disease-causing mutations [1]. For genetic researchers trying to unravel the disease-causing genes of over 6,800 rare diseases [2], exome sequencing enables the identification of common single-nucleotide variants (SNVs), copy number variations (CNVs), and small insertions or deletions (indels), as well as rare de novo mutations that may explain the heritability of Mendelian and complex disorders [3].
What are the benefits of exome sequencing?
  • Focused search for variants—almost 85% of disease-causing mutations are located in the exome
  • Lower costs—exome sequencing can be performed at a fraction of the cost of whole-genome sequencing
Learn more about exome sequencing >
Exome sequencing application note
Exome sequencing getting started
For Research Use Only. Not for use in diagnostic procedures.