Search Thermo Fisher Scientific
QuantStudio 3 and 5 real-tme PCR systems are factory-calibrated for quick installation and immediate use. These systems detect differences in target quantity as low as 1.5-fold, allow remote monitoring, and feature optional cloud-based Applied Biosystems analysis modules and data sharing.
Designed for both new and experienced users, they support 96-well (0.2ml or 0.1ml) or 384-well (QS5 only) consumables. The systems also feature multiplexing capabilities for up to 6 targets and software to enable 21 CFR Part 11 compliance.
This compact instrument can be configured as a standalone or with a computer to fit any laboratory needs.
Pre-optimized protocol templates minimize new user training
Applied Biosystems 96-well blocks feature VeriFlex technology with three or six independent temperature zones for precise temperature control and optimization.
Allows you to remotely monitor your runs, quickly analyze sophisticated data sets, store results in a secure space, and safely share them online
96-well (0.1-mL block), 96-well (0.2-mL block), and 384-well options
Security, auditing, e-signature package included
Maximize the performance of your QuantStudio 3 and 5 systems with the following calibration kits.
Category | Specification | QuantStudio 3 system | QuantStudio 5 system |
---|---|---|---|
Format |
Fixed blocks | ||
Efficiency and productivity |
Small footprint | ||
Multiplexing |
Up to 4 | 96-well up to 6 384-well up to 5 |
|
VeriFlex temperature control |
3 zones | 6 zones (96-well blocks only) | |
Direct connection to cloud-enabled system | |||
Full-function touchscreen | |||
On-board memory | Yes, >2K runs | Yes, >2K runs |
|
Features to assist with 21 CFR part 11 compliance |
Security | Security, auditing, e-signature package included | |
Dimensions |
40 x 27 x 50 cm (H x W x D) | 40 x 27 x 50 cm (H x W x D) | |
Maximize uptime |
Smart Help |
||
Minimal calibration for easy maintenance |
|||
Support |
On-site installation by professionally trained field service engineers (region dependent) |
||
SmartStart Orientation hands-on training package |
|||
Faster troubleshooting and resolution with Smart Remote Support tool |
For Research Use Only. Not for use in diagnostic procedures.