• Higher specificity–minimize primer-dimer and nonspecific amplification with a highly purified DNA polymerase with a proprietary hot start mechanism
  • Better performance–consistent amplification across a wide dynamic range
  • Lower price–priced to meet many lab budgets
  • Greener™ dye–for more brightness
  • Carry-over contamination control– 
contains heat-labile UDG
  • Broader compatibility–optimized for most real-time PCR instruments