Search Thermo Fisher Scientific
A mixture of live and dead bovine sperm cells stained with the dyes provided in our LIVE/DEAD® Sperm Viability Kit (Cat. no. L7011). Live sperm with intact membranes are labeled with our proprietary cell-permeant nucleic acid stain, SYBR® 14, and fluoresce green. Dead sperm, which have been killed by unprotected freeze-thawing, are labeled with propidium iodide (Cat. no. P1304MP, P3566, P21493) and fluoresce red-orange. The image was contributed by Duane L. Garner, School of Veterinary Medicine, University of Nevada, Reno, and Lawrence A. Johnson, USDA Agricultural Research Service.
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›