Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The CTS Xenon Electroporation System offers reliable delivery of DNA, RNA, proteins, and other molecules into cells, with exceptional cell viability and recovery. This leads to efficient transfection and genome editing, even in hard-to-transfect cells. A choice of two single-use electroporation chambers of different capacities provides flexibility: the CTS SingleShot chamber enables process development while the CTS MultiShot cartridge can be used in a closed, commercial-scale transfection system that helps minimize contamination and human error.
Process- and protocol-compatible with the Invitrogen Neon NxT Electroporation System, the CTS Xenon system enables scale-up from research to process development to commercial manufacturing with large-scale electroporation volumes of up to 25 mL. Pair it with other Thermo Fisher Scientific instruments to assemble a closed, integrated, GMP-compliant workflow that can be controlled by process automation software.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products.