Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Nitrated tyrosine residues in bovine pulmonary artery endothelial cells detected with rabbit anti-nitrotyrosine antibody. Fixed and permeabilized bovine pulmonary artery endothelial cells were treated with either degraded peroxynitrite (top panel) or 100 µM peroxynitrite (bottom panel) for five minutes at room temperature to induce protein nitration. Nitrated tyrosine residues were detected with our rabbit anti-nitrotyrosine polyclonal antibody (Cat. No. A21285) and visualized with the green-fluorescent Alexa Fluor® 488 goat anti–rabbit IgG antibody (Cat. No. A11008). Nuclei were counterstained with blue-fluorescent DAPI (Cat. No. D1306, D3571, D21490).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›