Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
In order to induce an influx of Ca2+, the cells were depolarized with 50 mM KCl in frame 2 and exposed to 5 µM ionomycin (I24222, a Ca2+ ionophore) in frame 8. The images are pseudocolored according to fluorescence intensity, with red representing high Ca2+ concentrations and blue representing low Ca2+ concentrations. The images were acquired using a fluorescence microscope equipped with a longpass filter set appropriate for fluorescein and a Photometrics Quantix cooled CCD camera.