Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
An orthogonal (x-z) display representing a microscope point-spread function. The microsphere used is a component of the PS-Speck™ Microscope Point Source Kit (Cat. no. P7220). This pseudocolored image was generated electronically from a series of microsphere images taken in x-y planes. The image was contributed by Jennifer Kramer, Scanalytics.
Images from confocal laser-scanning microscope optical cross-sectioning of our 15 µm FocalCheck™ microspheres that have a dark red–fluorescent ring stain with a green-fluorescent stain throughout the bead. Go ›
A mixture of blue fluorescent, red fluorescent and yellow-green fluorescent FluoSpheres® 4 µm sulfate microspheres. Go ›