Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Mini Gel Tank is designed for more intuitive use and greater convenience compared to traditional electrophoresis tanks. The unique, side-by-side tank design allows you to perform electrophoresis of 1 or 2 mini gels. The two separate gel chambers allow use of less running buffer, only use the buffer needed for each gel. The forward facing well configuration makes it easy to load samples. Each Mini Gel Tank undergoes a rigorous stress-relieving process that reduces tensions in the plastic to help prevent cracking. Made of durable polycarbonate and fit securely in a robust supporting base, the Mini Gel Tank is designed to stand the test of time. The Mini Gel Tank is compatible will all Invitrogen precast mini gels and SureCast handcast mini gels, and is also available in Protein Gels Welcome Packs. Optional Mini Blot Modules enable you to perform wet transfers in the same tank.
Capacity | Up to 2 mini gels |
Gel compatibility | Gel size: 8 x 8cm Gel cassette: 10 x 10cm Thickness: 1.0mm or 1.5mm |
Compatible gels | All Invitrogen precast mini gels, SureCast handcast gels, and Invitrogen empty mini gel cassettes |
Buffer chamber requirement | 400 mL for each mini gel |
Optional transfer apparatus | Mini Blot Module |
Transfer capacity | Up to 2 mini gels |
Compatible power supplies | PowerEase Touch 120W and PowerEase Touch 350W, Owl systems, for other systems such as Hoefer or Bio-Rad use with Novex Power Supply Adapters (Cat. No. ZA10001) |
Unit dimensions | 32 x 11.5 x 16 cm (height with lid) |
Material | Polycarbonate |
Electrode wire | Platinum |
Electrode limits | 500 VDC or 100 Watts |
Chemical resistance | The Mini Gel Tank is impervious to most alcohols but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene), or acetone. |
Catalog number | A25977 |
The Mini Gel Tank has a unique tank design that enables convenient side-by-side gel loading and enhanced viewing during use. Each tank can accommodate up to two gels per run where each gel is run in its own separate chamber- making it easy to run just one gel. This advantage allows for lower buffer requirements- only use the buffer needed to run one gel.
The Mini Gel Tank can be used for wet tank transfers using the Mini Blot Module. Two Mini Blot Modules fit into the Mini Gel Tank to allow transfer of two gels at a time. The unique design of the blot module allows for lower transfer buffer- only 220 mL per gel. This lower requirement keeps the amount of methanol needed to a minimum. Learn more about transferring in the Mini Gel Tank here.
Learn how to separate proteins using the Invitrogen Mini Gel Tank.
Learn how to perform a western wet tank transfer using the Invitrogen Mini Blot Module in the Mini Gel Tank.
Gel Type | Voltage (V) | Current (mA) | Ending Current | Run Time |
---|---|---|---|---|
Bolt Bis-Tris Plus 4-12% (MES) | 200 | 160 | 70 | 20 |
Bolt Bis-Tris Plus 4-12% (MOPS) | 200 | 160 | 50 | 35 |
NuPAGE 4-12% BisTris (MES) | 200 | 160 | 90 | 30 |
NuPAGE 4-12% BisTris (MOPS) | 200 | 140 | 50 | 42 |
Novex 4-20% TrisGlycine (denatured) | 125 | 40 | 10 | 100 |
Novex 4-20% TrisGlycine (native) | 125 | 30 | 10 | 90 |
NuPAGE 3-8% TrisAcetate (denatured) | 150 | 60 | 20 | 50 |
NuPAGE 3-8% TrisAcetate (native) | 150 | 40 | 10 | 100 |
Novex 10-20% Tricine | 125 | 110 | 40 | 65 |
NativePAGE 3-12% | 150 | 10 | <10 | 80 |
pH 3-10 IEF | program | 8 | 5 | 150 |
10% Zymogram (Gelatin) | 125 | 40 | 10 | 90 |
10% TBE-Urea (DNA) | 180 | 10 | 10 | 50 |
Capacity | Up to 2 mini gels |
Gel compatibility | Gel size: 8 x 8cm Gel cassette: 10 x 10cm Thickness: 1.0mm or 1.5mm |
Compatible gels | All Invitrogen precast mini gels, SureCast handcast gels, and Invitrogen empty mini gel cassettes |
Buffer chamber requirement | 400 mL for each mini gel |
Optional transfer apparatus | Mini Blot Module |
Transfer capacity | Up to 2 mini gels |
Compatible power supplies | PowerEase Touch 120W and PowerEase Touch 350W, Owl systems, for other systems such as Hoefer or Bio-Rad use with Novex Power Supply Adapters (Cat. No. ZA10001) |
Unit dimensions | 32 x 11.5 x 16 cm (height with lid) |
Material | Polycarbonate |
Electrode wire | Platinum |
Electrode limits | 500 VDC or 100 Watts |
Chemical resistance | The Mini Gel Tank is impervious to most alcohols but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene), or acetone. |
Catalog number | A25977 |
The Mini Gel Tank has a unique tank design that enables convenient side-by-side gel loading and enhanced viewing during use. Each tank can accommodate up to two gels per run where each gel is run in its own separate chamber- making it easy to run just one gel. This advantage allows for lower buffer requirements- only use the buffer needed to run one gel.
The Mini Gel Tank can be used for wet tank transfers using the Mini Blot Module. Two Mini Blot Modules fit into the Mini Gel Tank to allow transfer of two gels at a time. The unique design of the blot module allows for lower transfer buffer- only 220 mL per gel. This lower requirement keeps the amount of methanol needed to a minimum. Learn more about transferring in the Mini Gel Tank here.
Learn how to separate proteins using the Invitrogen Mini Gel Tank.
Learn how to perform a western wet tank transfer using the Invitrogen Mini Blot Module in the Mini Gel Tank.
Gel Type | Voltage (V) | Current (mA) | Ending Current | Run Time |
---|---|---|---|---|
Bolt Bis-Tris Plus 4-12% (MES) | 200 | 160 | 70 | 20 |
Bolt Bis-Tris Plus 4-12% (MOPS) | 200 | 160 | 50 | 35 |
NuPAGE 4-12% BisTris (MES) | 200 | 160 | 90 | 30 |
NuPAGE 4-12% BisTris (MOPS) | 200 | 140 | 50 | 42 |
Novex 4-20% TrisGlycine (denatured) | 125 | 40 | 10 | 100 |
Novex 4-20% TrisGlycine (native) | 125 | 30 | 10 | 90 |
NuPAGE 3-8% TrisAcetate (denatured) | 150 | 60 | 20 | 50 |
NuPAGE 3-8% TrisAcetate (native) | 150 | 40 | 10 | 100 |
Novex 10-20% Tricine | 125 | 110 | 40 | 65 |
NativePAGE 3-12% | 150 | 10 | <10 | 80 |
pH 3-10 IEF | program | 8 | 5 | 150 |
10% Zymogram (Gelatin) | 125 | 40 | 10 | 90 |
10% TBE-Urea (DNA) | 180 | 10 | 10 | 50 |
For Research Use Only. Not for use in diagnostic procedures.