Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Scientific dialysis devices are available in a variety of dialysis options for the rapid and trouble-free dialysis of sample volumes ranging from 10 μL–250 mL. Unlike traditional dialysis tubing, these innovative devices do not require knots or clips that can lead to leaks and sample loss.
Pierce 96-well Microdialysis Plates and Slide-A-Lyzer Dialysis MINI Devices are excellent for small volumes, Slide-A-Lyzer G3 Dialysis Cassettes are recommended for small to medium volumes, and Slide-A-Lyzer Dialysis Flasks are recommended for larger volumes.
Dialysis product | Traditional tubing | Cassette |
Sample handling | Slippery when wet Flat tubing is difficult to handle when wet, increasing risk of sample loss during sample addition/removal | Easy handling and secure sample delivery Plastic frame and cap allow for secure sample addition/removal |
Sample recovery | Risk of sample loss Sample can leak out if clamp becomes loose or falls off | Sample protection Welded membrane and leak-proof cap helps minimize the risk of sample loss |
Speed | Time consuming Non-optimized design that typically requires overnight dialysis | Fast and efficient Innovative design with high surface/volume ratio allows for more rapid dialysis than conventional tubing |
Learn more | Learn more |
Slide-A-Lyzer Dialysis Devices | Microdialysis Plates | SnakeSkin Dialysis Tubing | |||
Slide-A-Lyzer Cassettes | Slide-A-Lyzer Flasks | Slide-A-Lyzer MINI Dialysis Devices | |||
External float required? | No | Yes | Yes (0.1 mL size only) | No | No |
Method of sample addition/removal | Pipette | Pipette, Directly pouring | Pipette | Pipette | Pipette |
Volume range | 1–125 mL | 150–250 mL | 10 μL–2 mL | 10–100 μL | 10–100mL |
Number of devices with different volume capacities | 4–5 | 1 | 3 | 1 | 3 |
Molecular weight cutoffs (MWCOs) | 2K, 3.5K, 7K (original only), 10K, 20K | 2K, 3.5K, 10K, 20K | 2K (0.1 mL size only), 3.5K, 10K, 20K | 3.5K, 10K | 3.5K, 7K, 10K |
Automation compatible? | No | No | No | Yes | No |
Integrated buffer reservoir? | No | No | Yes, except 0.1 mL size | Yes | No |
Gamma-irradiated options? | No, E-Beam irradiated available (G3 device) Yes (original device) | No | No | No | No |
Molecular weight cutoff (MWCO) color coded? | Yes | Yes | Yes | No | Yes |
Find products | Find products | Find products |
For Research Use Only. Not for use in diagnostic procedures.