Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Transcriptome sequencing, or RNA sequencing (RNA-Seq), provides fundamental insights into how genomes are organized and regulated—giving us valuable information about the internal state of cells and how altered expression of genetic variants contributes to complex disease research. Transcriptome sequencing relies on next-generation sequencing (NGS) methodology and techniques.
We offer two great options for transcriptome analysis:
Learn more about the Ion AmpliSeq Transcriptome solution ›
Ion AmpliSeq Transcriptome Human Gene Expression Kit application note
Exosome miRNA sequencing for biomarker profiling using Ion S5 System and Ion Total RNA-Seq Kit
Lesley Cheng, PhD, La Trobe University.
We offer a complete RNA sequencing solution for gene expression research. From sample prep to analysis, we enable you to discover more when you perform RNA sequencing. We also support verification of your findings with our qPCR systems and reagents.
Our whole transcriptome RNA sequencing workflow provides you with:
The Invitrogen RiboMinus Eukaryote Kit v2 provides more than 90% rRNA depletion, which helps increase the sequencing power of the Ion Proton System
The Ion Total RNA-Seq Kit v2 provides a fast, simple workflow with as little as 100 ng of total RNA input, while maintaining strand orientation and minimizing bias and error.
Use the Ion Chef System for walk-away template preparation and chip loading. Suitable for users of all experience levels.
The Ion GeneStudio S5 systems are ideal for sequencing of gene panels all the way to transcriptomes. The sequencing run time is as little as 2.5 hours, with only 15 minutes of setup time.
Torrent Suite Software automatically provides the sequence reads in exportable FASTQ or BAM formats, which can be uploaded to third-party software, such as Partek Flow software packages
The Invitrogen RiboMinus Eukaryote Kit v2 provides more than 90% rRNA depletion, which helps increase the sequencing power of the Ion Proton System
The Ion Total RNA-Seq Kit v2 provides a fast, simple workflow with as little as 100 ng of total RNA input, while maintaining strand orientation and minimizing bias and error.
Use the Ion Chef System for walk-away template preparation and chip loading. Suitable for users of all experience levels.
The Ion GeneStudio S5 systems are ideal for sequencing of gene panels all the way to transcriptomes. The sequencing run time is as little as 2.5 hours, with only 15 minutes of setup time.
Torrent Suite Software automatically provides the sequence reads in exportable FASTQ or BAM formats, which can be uploaded to third-party software, such as Partek Flow software packages
Ion Torrent semiconductor sequencing combines the simple, integrated wet-lab workflow with Torrent Suite Software and third-party solutions for fast identification, characterization, and reporting of the most biologically interesting transcripts.
Torrent Suite Software provides you the tools that take you from raw sequence data to informative results, including optimized signal processing, base calling, transcriptome analysis, sequence alignment, and variant analysis. Post-run sequencing data are available for download with a simple right-click. Sequencing run reports are easily browsed, with expandable analysis plots and straightforward tables summarizing key results to help ensure that sequencing runs are of high quality.
Transcriptome-specific workflows in third-party software such as Partek™ Flow™ software are available to perform read alignment, transcript level abundance measurements, and multi-sample differential gene expression analysis. Additionally, quality control metrics and interactive data visualization is provided.