새로운 HPLC 시스템에 대한 투자를 생각해보십시오.
HPLC 시스템 총 구입 및 유지 비용 계산기는 지정된 기간 동안 Thermo Scientific HPLC 시스템을 구매하고 운영하는데 드는 총 비용을 추정하는데 도움이 됩니다. 다른 기기와 비교하여 최고의 제품을 찾으십시오. 
현재 설정과 관련된 일반적인 시스템 사용 조건 및 일반 소모품 비용을 입력하여 시료당 예상 비용과 함께 연간 실행 가능한 시료 개수를 추정하십시오. 탠덤 및 듀얼 LC 워크플로우의 적용이 어떻게 생산성을 높이고 시료당 비용을 절감하여 보다 빠른 투자 회수가 가능한지 확인하십시오.
사용 책임은 사용자에게 있습니다. 미리 채워진 데이터는 추정치입니다. Thermo Fisher Scientific은 데이터, 계산 또는 결과의 정확성에 대해 책임을 지지 않습니다.
Step 1: Choose a preset process OR start with minimum conditionals. After, enter your own adjustments.
Minimum conditionals
or
Preset process
Additional experiment conditions
Format: 1000; not 1,000.00
Not required for Isocratic conditions
Step 2: Review the results
Step 3: Review the charts
Number of Systems Required to Achieve
Target Number of Samples Per Year
Single
Tandem
Dual
Number of Samples Per Day
Step 4: Compare
Most accurate results require content in these fields
Single Channel LCTandem LC or LC-MSDual LC
Click to enlarge
Click to enlarge
Click to enlarge
System Cost ($)
Yearly Maintenance Costs ($)
Yearly Labor and Overhead Costs ($)
Additional Cost Per Sample ($)
System Type
Cost Per Sample ($)
Based on the maximum number of samples
Number of Samples Per Day Per System
Number of Samples Per Year Per System
Yearly System, Maintenance, Overhead Costs ($)
Yearly Column and Consumable Costs ($)
Yearly Solvent Costs ($)
System ASystem B
System Cost ($)
Yearly Maintenance Costs ($)
Yearly Labor and Overhead Costs ($)
Additional Cost Per Sample ($)
System Type
Cost Per Sample ($)
Based on the maximum number of samples
Number of Samples Per Day Per System
Number of Samples Per Year Per System
Yearly System, Maintenance, Overhead Costs ($)
Yearly Column and Consumable Costs ($)
Yearly Solvent Costs ($)
Step 5: Choose product
Vanquish Core HPLC SystemsVanquish Flex UHPLC SystemsVanquish Horizon UHPLC SystemsVanquish Neo UHPLC System
General application
Dependable routine HPLC analysisFlexible HPLC and UHPLC method developmentTop performance and resolution for demanding UHPLC separationsMaximum low-flow performance for high-sensitivity LC-MS workflows
Pump options
Binary, Quaternary, Dual and IsocraticBinary, Quaternary and DualBinaryBinary
System pressure
Up to 700 barUp to 1000 barUp to 1500 barUp to 1500 bar
Flow range
0.001 – 10 mL/min0.001 – 8 mL/min0.001 – 5 mL/min 1 nL/min – 100 µL/min
Biocompatible
NoYesYesYes
Common detector setup
Up to 2 detectors and/or 1 mass spectrometerUp to 2 detectors and/or 1 mass spectrometerUp to 2 detectors and/or 1 mass spectrometer1 VWD and/or 1 mass spectrometer
Vanquish Duo UHPLC Systems for Dual LCVanquish Duo UHPLC Systems for Dual LC-MSVanquish Duo HPLC and UHPLC Systems for Tandem LC or LC-MS
Benefit
Doubled productivity with 2 independent UHPLC channels built into one stackIntegrated multi-channel LC for ultra-high-throughput LC-MSMaximized detector utilization by using offline column reconditioning
Supported systems
Vanquish Flex
Vanquish Horizon
Vanquish Flex
Vanquish Horizon
Vanquish Core
Vanquish Flex
Vanquish Horizon
Common detector setup
1-2 detectors per channel
(2-4 detectors per system)
1 mass spectrometerUp to 2 detectors and/or 1 mass spectrometer
Software control
Thermo Scientific Chromeleon CDS
Thermo Scientific Standard System
Integration for Waters™ Empower™ 3 CDS software
Thermo Scientific Aria MX softwareThermo Scientific Chromeleon CDS
Thermo Scientific Standard System
Integration for Thermo Scientific XCalibur software
Vanquish Flex Duo Ternary UHPLC System with 2 Detectors
Pump type:DPG (Vanquish Dual Pump F)
Maximum pressure:1000 bar, 15,000 psi
Autosampler:Dual (Vanquish Dual Split Sampler FT)
Column switching:N/A
Detectors required:2 x Vanquish Detectors and/or Mass Spectrometer
Vanquish Flex Duo Binary UHPLC System with 2 Detectors
Pump type:2 x HPG (Vanquish Binary Pump F)
Maximum pressure:1000 bar, 15,000 psi
Autosampler:Dual (Vanquish Dual Split Sampler FT)
Column switching:N/A
Detectors required:2 x Vanquish Detectors and/or Mass Spectrometer
Vanquish Horizon Duo Binary UHPLC System with 2 Detectors
Pump type:2 x HPG (Vanquish Binary Pump H)
Maximum pressure:1500 bar, 22,000 psi
Autosampler:Dual (Vanquish Dual Split Sampler HT)
Column switching:N/A
Detectors required:2 x Vanquish Detectors and/or Mass Spectrometer