Search Thermo Fisher Scientific
Invitrogen Lipofectamine RNAiMAX Transfection Reagent, our best siRNA transfection reagent, offers an advanced, efficient solution for siRNA delivery. No other siRNA-specific transfection reagent provides such easy and efficient siRNA delivery in a wide variety of cell lines. You can achieve the highest transfection efficiencies in cell lines including the common cell types, stem cells and primary cells, as well as traditionally hard-to-transfect cell types.
When it comes to achieving effective gene knockdown, Lipofectamine RNAiMAX Transfection Reagent outperforms other siRNA transfection reagents. High knockdown levels of target genes can be achieved with as little as 1 nM siRNA (Figure 1). The Lipofectamine RNAiMAX protocol recommends using 10 nM siRNA as a starting point to maximize knockdown levels. Even when coupled with low concentrations of siRNA, RNAiMAX reagent helps obtain high knockdown levels while is also beneficial in minimizing background expression from untransfected cells and to minimize other non-specific effects.
Figure 1. Superior knockdown with Lipofectamine RNAiMAX Transfection Reagent compared to competing siRNA transfection reagents. At both 10 nM and 1 nM p53 siRNA, Lipofectamine RNAiMAX Transfection Reagent provides more effective knockdown relative to non-transfected cells than other RNAi reagents, including siLentFect™ (Bio-Rad), DharmaFECT™ (Dharmacon), and HiPerFect (Qiagen) reagents.
Transfection-mediated cytotoxicity can mask the true phenotype of a target gene being studied, so minimizing the amount of reagent used in your transfections is a critical factor for successful RNAi experiments.
Lipofectamine RNAiMAX Transfection Reagent gives maximal knockdown and excellent cell viability across a 10-fold concentration range of the reagent (Figure 2). This broad peak of optimal transfection activity with minimal cytotoxicity means you can be assured of achieving high knockdown levels despite differences in cell density, minor experimental inaccuracies, and other variations. Not also does this make it easy to optimize Lipofectamine RNAiMAX reagent for the lowest siRNA concentration but also reduces cellular stress and cytotoxicity in your experimental system simultaneously.
Figure 2. Optimal knockdown and minimal cytotoxicity in A549 cells transfected with Lipofectamine RNAiMAX Transfection Reagent. A range of 0.1 μL to 1.0 μL of Lipofectamine RNAiMAX reagent was used, resulting in efficient knockdown of the p53 gene expression level while maintaining excellent cell viability.
Lipofectamine RNAiMAX offers a simple protocol to achieve high transfection efficiencies in many cell types. There is no need to remove Lipofectamine-siRNA complexes, or change or add cell culture media following transfection.
When combined with Invitrogen BLOCK-iT Alexa Fluor Red Fluorescent Control, Lipofectamine RNAiMAX reagent gives you the most versatile approach to all your knockdown experiments (Table 1, Figure 3). Spend less hands-on time and get better results faster.
Figure 3. Improve your transfection efficiencies using the BLOCK-iT Alexa Fluor Red Fluorescent Control. (A) HeLa cells were transfected with the BLOCK-iT Alexa Fluor Red Fluorescent Control (50 nM) using Lipofectamine RNAiMAX Transfection Reagent. (B) 24 hours post-transfection, nearly 100% of the cells take up the control and retain a normal morphology, as seen in the bright-field image.
Cell line/type | Cell type |
---|---|
MDA-MB-435 | Breast cancer |
HeLa | Cervical carcinoma |
HT1080 | Human fibrosarcoma |
A549 | Lung carcinoma |
HepG2 | Liver carcinoma |
MCF7 | Breast cancer |
SK-N-SH | Neuroblastoma |
Mesenchymal stem cells | Bone marrow |
HAMSC | Human aortic smooth muscle cells |
HEKa | Human epidermal keratinocytes |
HRE | Human renal epithelial cells |
NTERA-2 | Embryonal carcinoma cells |
HUVEC | Human umbilical vein endothelial cells |
ADSC | Adipose-derived stem cells |
HCT116 | Colon carcinoma |
Follow this step-by-step video to perform a successful siRNA transfection using Lipofectamine RNAiMAX reagent.
Packaging must protect the product inside—but that doesn’t have to mean sacrificing sustainability. Lipofectamine products are shipped at ambient temperature, diverting 24,000 ft3 of Expanded Polystyrene (EPS) coolers from landfills annually. We reduce the carbon footprint from packaging and shipping these products by 80 tons (CO2 equivalents).
Learn more about sustainable solutions
Download the green fact sheet
Our Lipofectamine reagent-specific protocols have been optimized for efficiency, viability, and reproducibility across a broad range of cell types. Search this library to find protocols and citations curated to fit your experimental needs. Filter by cell type, payload, and product of choice to easily unlock your answers.
Search our extensive collection of Transfection FAQs to find the answers related to Lipofectamine RNAiMAX reagent.