Search Thermo Fisher Scientific
Our reliable and easy to use Thermo Scientific™ Wellwash™ microplate strip washers provide secure washing performance for routine and research ELISA applications. Wellwash washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease-of-use, convenience, and safety.
Wellwash | Wellwash Versa | |
Wash head | 1x8, 1x12 | two 1x8, two 1x12, 2x8 cell wash head |
---|---|---|
Wash bottles | 1 bottle (2 liter) | 3 bottles (two 4 liter and one 2 liter) |
Plate formats | 96 | 96, 384 |
Cell washing | No | Yes |
Robot compatibility | No | Yes |