Search Thermo Fisher Scientific
Selecting the correct vacuum concentrator system is critical for optimum performance and reliability. Savant SpeedVac Kits come in two formats:
|
|
Use the charts below to select the right system for your application. Email speedvac@thermofisher.com for any ordering questions or to discuss a customized system to suit your needs.
SpeedVac Models* | DNA130 | SPD1030P1 with | SRF110P1/SRF110P2 | SPD120P1 | SPD120P2 | SPD1030 | SPD130P1 | SPD140P1 | SPD140P2 |
---|---|---|---|---|---|---|---|---|---|
Format | Integrated | Integrated | Modular | Modular | Modular | Integrated | Modular | Modular | Modular |
Max 1.5mL tube capacity | 36 | 120 | 132 | 120 | 120 | 120 | 120 | 120 | 120 |
DNA/RNA Ethanol, H2O, buffers | |||||||||
DNA/RNA Ammonium hydroxide (oligos) | |||||||||
DNA/RNA DNA in acetonitrile or H2O | |||||||||
Proteins, enzymes, HPLC fractions H20, acetonitrile, methanol, ethanol + .1% TFA | |||||||||
Low boiling-point solvents (Aggressive) organic chemistry, drug discovery Chloroform, ethyl acetate, acetone, methylene chloride, hexane | |||||||||
High boiling point solvents (Aggressive) organic chemistry, drug discovery DMSO, DMF, toluene, nitrobenzene, pyridine | |||||||||
Strong acids and bases Organic chemistry, drug discovery TFA, |
*All kits linked to 115V/60Hz models. 230/50Hz models also available on relevant product pages.
SpeedVac Models* | SPD1030 | SPD2030P1 with | SPD210P1 | SPD210P2 | SPD2030 | SPD300P2 | SPD300P1 |
---|---|---|---|---|---|---|---|
Format | Integrated | Integrated | Modular | Modular | Integrated | Modular | Modular |
Max 1.5mL tube capacity | 120 | 200 | 200 | 200 | 200 | 200 | 200 |
DNA/RNA Ethanol, H2O, buffers | |||||||
DNA/RNA Ammonium hydroxide (oligos) | |||||||
DNA/RNA DNA in acetonitrile or H2O | |||||||
Proteins, enzymes, HPLC fractions H20, acetonitrile, methanol, ethanol + .1% TFA | |||||||
Low boiling-point solvents (Aggressive) organic chemistry, drug discovery Chloroform, ethyl acetate, acetone, methylene chloride, hexane | |||||||
High boiling point solvents (Aggressive) organic chemistry, drug discovery DMSO, DMF, toluene, nitrobenzene, pyridine | |||||||
Strong acids and bases Organic chemistry, drug discovery TFA, HCL, acetic acid, formic acid, sodium hydroxide |
*All kits linked to 115V/60Hz models if available. 230V/50Hz models are also available on the relevant product pages
SpeedVac Models* | DNA130 | SPD1030P1 with | SRF110P1/SRF110P2 | SPD120P1 | SPD120P2 | SPD1030 | SPD130P1 | SPD140P1 | SPD140P2 |
---|---|---|---|---|---|---|---|---|---|
Format | Integrated | Integrated | Modular | Modular | Modular | Integrated | Modular | Modular | Modular |
Max 1.5mL tube capacity | 36 | 120 | 132 | 120 | 120 | 120 | 120 | 120 | 120 |
DNA/RNA Ethanol, H2O, buffers | |||||||||
DNA/RNA Ammonium hydroxide (oligos) | |||||||||
DNA/RNA DNA in acetonitrile or H2O | |||||||||
Proteins, enzymes, HPLC fractions H20, acetonitrile, methanol, ethanol + .1% TFA | |||||||||
Low boiling-point solvents (Aggressive) organic chemistry, drug discovery Chloroform, ethyl acetate, acetone, methylene chloride, hexane | |||||||||
High boiling point solvents (Aggressive) organic chemistry, drug discovery DMSO, DMF, toluene, nitrobenzene, pyridine | |||||||||
Strong acids and bases Organic chemistry, drug discovery TFA, |
*All kits linked to 115V/60Hz models. 230/50Hz models also available on relevant product pages.
SpeedVac Models* | SPD1030 | SPD2030P1 with | SPD210P1 | SPD210P2 | SPD2030 | SPD300P2 | SPD300P1 |
---|---|---|---|---|---|---|---|
Format | Integrated | Integrated | Modular | Modular | Integrated | Modular | Modular |
Max 1.5mL tube capacity | 120 | 200 | 200 | 200 | 200 | 200 | 200 |
DNA/RNA Ethanol, H2O, buffers | |||||||
DNA/RNA Ammonium hydroxide (oligos) | |||||||
DNA/RNA DNA in acetonitrile or H2O | |||||||
Proteins, enzymes, HPLC fractions H20, acetonitrile, methanol, ethanol + .1% TFA | |||||||
Low boiling-point solvents (Aggressive) organic chemistry, drug discovery Chloroform, ethyl acetate, acetone, methylene chloride, hexane | |||||||
High boiling point solvents (Aggressive) organic chemistry, drug discovery DMSO, DMF, toluene, nitrobenzene, pyridine | |||||||
Strong acids and bases Organic chemistry, drug discovery TFA, HCL, acetic acid, formic acid, sodium hydroxide |
*All kits linked to 115V/60Hz models if available. 230V/50Hz models are also available on the relevant product pages