View the relevant questions below:
Beginning your experiment? Visit our
Here are some suggestions:
Here are some suggestions:
Pre-stained standards have a dye that is covalently bound to each protein that will result in the standard migrating differently in different buffer systems (i.e., different gels). As a result, using a pre-stained standard for molecular weight estimation will only give the apparent molecular weight of the protein. Pre-stained standards may be used for molecular weight approximation, confirming gel migration and estimating blotting efficiency but for accurate molecular weight estimation, an unstained standard should be used.
The fading is most likely due to detergent in the western blocking/washing solutions that can remove some of the proteins from the membrane. The dye itself will not wash off of the proteins because it is covalently bound. We have found that smaller pore size membranes retain the proteins better during blocking and wash procedures, and hence recommend use of 0.2 μm instead of 0.45 μm membranes for best resolution and protein retention. After transfer, it is a good idea to circle the pre-stained bands with a pencil on the membrane, so band positions can be identified after blocking and processing.
Here are some suggestions:
Here are possible causes and solutions:
Cause | Solution |
Sample was boiled | Discard boiled aliquot. |
Too much volume of ladder used | Add less volume or dilute the ladder in protein loading buffer prior to use. |
Here are possible causes and solutions:
Cause | Solution |
Not enough volume of ladder loaded on the gel | Load an appropriate volume of the ladder onto the gel. Here are our recommendations:
|
Incomplete or poor transfer | Optimize transfer conditions |
Here are possible causes and solutions:
Cause | Solution |
Sample was boiled | Discard boiled aliquot. |
Too much volume of ladder used | Add less volume or dilute the ladder in protein loading buffer prior to use. |
Here are possible causes and solutions:
Cause | Solution |
Not enough volume of ladder loaded on the gel | Load an appropriate volume of the ladder onto the gel. Here are our recommendations:
|
Incomplete or poor transfer | Optimize transfer conditions |
Here are possible causes and solutions:
Cause | Solution |
Sample was boiled | Discard boiled aliquot. |
Too much volume of ladder used | Add less volume or dilute the ladder in protein loading buffer prior to use. |
Here are possible causes and solutions:
Cause | Solution |
Not enough volume of ladder loaded on the gel | Load an appropriate volume of the ladder onto the gel. Here are our recommendations:
|
Incomplete or poor transfer | Optimize transfer conditions |
Here are possible causes and solutions:
Cause | Solution |
Sample was boiled | Discard boiled aliquot. |
Too much volume of ladder used | Add less volume or dilute the ladder in protein loading buffer prior to use. |
DTT oxidation in storage buffer | Add freshly prepared DTT solution to a final concentration of 100 mM. |
Here are possible causes and solutions:
Cause | Solution |
Sample was boiled | Discard boiled aliquot. |
Too much volume of ladder used | Add less volume or dilute the ladder in protein loading buffer prior to use. |
DTT oxidation in storage buffer | Add freshly prepared DTT solution to a final concentration of 50 mM. Heat for 10 minutes at 95 degrees C. Cool at room temperature and mix well. Store at –20 degrees C for further use. |
Here are possible causes and solutions:
Cause | Solution |
Not enough volume of ladder loaded on the gel | Load an appropriate volume of the ladder onto the gel. Here are our recommendations:
|
Incomplete or poor transfer | Optimize transfer conditions |
Here are possible causes and solutions:
Cause | Solution |
Sample was boiled | Discard boiled aliquot. |
Too much volume of ladder used | Load less volume of the ladder. |
Concentration of antibody is too high | Optimize antibody concentration |
Here are possible causes and solutions:
Cause | Solution |
Not enough volume loaded | Load more volume onto the gel |
Incomplete or poor transfer | Optimize transfer conditions |
Secondary antibody does not recognize marker proteins | Ensure appropriate secondary is used that binds to marker proteins. |
Secondary antibody not enzyme-conjugated | Ensure appropriate secondary antibody used in system |
A mouse monoclonal primary antibody was used | Use a greater volume of the ladder or use SuperSignal Enhanced Molecular Weight Protein Ladder (Cat. No. 84786) for mouse primary antibodies |
This is likely due to the dye-front not run off the gel or removed from the blot. We recommend running the dye front off of the gel or removing it from the blot.
Here are possible causes and solutions:
Cause | Solution |
Not enough volume loaded | Load more volume onto the gel |
Incomplete or poor transfer | Optimize transfer conditions |
Secondary antibody does not recognize marker proteins | Ensure appropriate secondary is used that binds to marker proteins. |
Secondary antibody not enzyme-conjugated | Ensure appropriate secondary antibody used in system |
Primary antibodies with low starting concentrations may result in insufficient chemiluminescent detection of the western blot positive control bands. If the unstained 30 kDa and 80 kDa bands produce weak or no signal, spike the diluted primary antibody with the corresponding rabbit IgG or mouse IgG to a concentration of 1-5 µg/mL, prior to secondary antibody incubation. Follow with respective secondary (GAM/GAR) incubation to increase the intensity of western blot positive control bands in the iBright Prestained Protein Ladder.