Search Thermo Fisher Scientific
Thermo Scientific continuous air particulate monitoring systems and manual ambient air particulate sampling instruments continuously and reliably monitor particulates in ambient air using advanced technology.
Use this product comparison guide to see which system is right for you.
TEOM™ 1405-DF | TEOM™ 1405-D | TEOM™ 1405-F | TEOM™ 1405 | ||||
![]() | ![]() | ![]() | ![]() | ||||
Measurement Method | |||||||
TEOM (Tapered Element Oscillating Microbalance) | ✓ | ✓ | ✓ | ✓ | |||
BETA Attenuation | |||||||
Hybrid (Beta Attenuation + Light Scattering) | |||||||
Gravimetric Filter Collection | |||||||
Operation Mode | |||||||
Continuous Measurement | ✓ | ✓ | ✓ | ✓ | |||
Manual Sampler | |||||||
Single Channel | ✓ | ✓ | |||||
Dual Channel | ✓ | ✓ | |||||
Regulatory Approval | |||||||
FRM US EPA PM-2.5 |
|
| |||||
FRM US EPA PM-10 |
|
| |||||
FEM US EPA PM-2.5 | ✓ | ✓ | |||||
FEM US EPA PM-10 | ✓ | ✓ | |||||
FEM US EPA PM-COARSE | ✓ | ||||||
TUV PM-2.5 | ✓ | ✓ | |||||
TUV PM-10 | ✓ | ✓ | |||||
China CCEP PM10 | ✓ | ||||||
China CCEP PM2.5 | ✓ |
Partisol™ 2025i | Partisol™ 2025i-D | Partisol™ 2000i | Partisol™ 2000i-D | ||||
![]() | ![]() | ![]() | ![]() | ||||
Measurement Method | |||||||
TEOM (Tapered Element Oscillating Microbalance) | |||||||
BETA Attenuation | |||||||
Hybrid (Beta Attenuation + Light Scattering) | |||||||
Gravimetric Filter Collection | ✓ | ✓ | ✓ | ✓ | |||
Operation Mode | |||||||
Continuous Measurement | |||||||
Manual Sampler | ✓ | ✓ | ✓ | ✓ | |||
Single Channel | ✓ | ✓ | |||||
Dual Channel | ✓ | ✓ | |||||
Regulatory Approval | |||||||
US EPA PM-2.5 | ✓ | ✓ | ✓ | ✓ | |||
US EPA PM-10 | ✓ | ✓ | ✓ | ✓ | |||
FEM US EPA PM-2.5 | |||||||
FEM US EPA PM-10 | |||||||
FEM US EPA PM-COARSE | ✓ | ✓ | |||||
TUV PM-2.5 | |||||||
TUV PM-10 | ✓ | ✓ | |||||
China CCEP PM10 | |||||||
China CCEP PM2.5 | ✓ | ✓ |
5014i Beta | 5028i | 5030i SHARP | ||||
![]() | ![]() | ![]() | ||||
Measurement Method | ||||||
TEOM (Tapered Element Oscillating Microbalance) | ||||||
BETA Attenuation | ✓ | ✓ | ||||
Hybrid (Beta Attenuation + Light Scattering) | ✓ | |||||
Gravimetric Filter Collection | ||||||
Operation Mode | ||||||
Continuous Measurement | ✓ | ✓ | ✓ | |||
Manual Sampler | ||||||
Single Channel | ✓ | ✓ | ||||
Dual Channel | ✓ | |||||
Regulatory Approval | ||||||
US EPA PM-2.5 |
|
|
| |||
US EPA PM-10 |
|
| ||||
FEM US EPA PM-2.5 | ✓ | ✓ | ✓ | |||
FEM US EPA PM-10 | ✓ | ✓ | ||||
FEM US EPA PM-COARSE | ||||||
TUV PM-2.5 | ✓ | ✓ | ||||
TUV PM-10 | ✓ | ✓ | ||||
China CCEP PM10 | ✓ | ✓ | ✓ | |||
China CCEP PM2.5 | ✓ | ✓ | ✓ |
TEOM™ 1405-DF | TEOM™ 1405-D | TEOM™ 1405-F | TEOM™ 1405 | ||||
![]() | ![]() | ![]() | ![]() | ||||
Measurement Method | |||||||
TEOM (Tapered Element Oscillating Microbalance) | ✓ | ✓ | ✓ | ✓ | |||
BETA Attenuation | |||||||
Hybrid (Beta Attenuation + Light Scattering) | |||||||
Gravimetric Filter Collection | |||||||
Operation Mode | |||||||
Continuous Measurement | ✓ | ✓ | ✓ | ✓ | |||
Manual Sampler | |||||||
Single Channel | ✓ | ✓ | |||||
Dual Channel | ✓ | ✓ | |||||
Regulatory Approval | |||||||
FRM US EPA PM-2.5 |
|
| |||||
FRM US EPA PM-10 |
|
| |||||
FEM US EPA PM-2.5 | ✓ | ✓ | |||||
FEM US EPA PM-10 | ✓ | ✓ | |||||
FEM US EPA PM-COARSE | ✓ | ||||||
TUV PM-2.5 | ✓ | ✓ | |||||
TUV PM-10 | ✓ | ✓ | |||||
China CCEP PM10 | ✓ | ||||||
China CCEP PM2.5 | ✓ |
Partisol™ 2025i | Partisol™ 2025i-D | Partisol™ 2000i | Partisol™ 2000i-D | ||||
![]() | ![]() | ![]() | ![]() | ||||
Measurement Method | |||||||
TEOM (Tapered Element Oscillating Microbalance) | |||||||
BETA Attenuation | |||||||
Hybrid (Beta Attenuation + Light Scattering) | |||||||
Gravimetric Filter Collection | ✓ | ✓ | ✓ | ✓ | |||
Operation Mode | |||||||
Continuous Measurement | |||||||
Manual Sampler | ✓ | ✓ | ✓ | ✓ | |||
Single Channel | ✓ | ✓ | |||||
Dual Channel | ✓ | ✓ | |||||
Regulatory Approval | |||||||
US EPA PM-2.5 | ✓ | ✓ | ✓ | ✓ | |||
US EPA PM-10 | ✓ | ✓ | ✓ | ✓ | |||
FEM US EPA PM-2.5 | |||||||
FEM US EPA PM-10 | |||||||
FEM US EPA PM-COARSE | ✓ | ✓ | |||||
TUV PM-2.5 | |||||||
TUV PM-10 | ✓ | ✓ | |||||
China CCEP PM10 | |||||||
China CCEP PM2.5 | ✓ | ✓ |
5014i Beta | 5028i | 5030i SHARP | ||||
![]() | ![]() | ![]() | ||||
Measurement Method | ||||||
TEOM (Tapered Element Oscillating Microbalance) | ||||||
BETA Attenuation | ✓ | ✓ | ||||
Hybrid (Beta Attenuation + Light Scattering) | ✓ | |||||
Gravimetric Filter Collection | ||||||
Operation Mode | ||||||
Continuous Measurement | ✓ | ✓ | ✓ | |||
Manual Sampler | ||||||
Single Channel | ✓ | ✓ | ||||
Dual Channel | ✓ | |||||
Regulatory Approval | ||||||
US EPA PM-2.5 |
|
|
| |||
US EPA PM-10 |
|
| ||||
FEM US EPA PM-2.5 | ✓ | ✓ | ✓ | |||
FEM US EPA PM-10 | ✓ | ✓ | ||||
FEM US EPA PM-COARSE | ||||||
TUV PM-2.5 | ✓ | ✓ | ||||
TUV PM-10 | ✓ | ✓ | ||||
China CCEP PM10 | ✓ | ✓ | ✓ | |||
China CCEP PM2.5 | ✓ | ✓ | ✓ |