Search Thermo Fisher Scientific
With 75 years of trusted single-use bioprocessing containment solutions, the Nalgene portfolio is designed with precision and innovation. Our products provide excellent containment and protection for your valuable biopharmaceutical processes.
We offer a comprehensive product portfolio of single-use bioprocessing solutions, so you can choose the products that meet your specific needs and budget. Rest assured in our product features and the peace of mind that comes from using a single supplier with a global supply chain.
Nalgene production bottles and carboys offer leakproof* storage for sensitive liquids, buffers, media, and more. They are available in PC, PETG, PP, LDPE, HDPE, FEP, and PFA in volumes ranging from 5 mL to 50 L. Extensive regulatory documentation helps ensure that the products meet your demanding biotechnology requirements and can easily integrate into your processes.
Application |
Standard |
Low particulate |
Ultralow particulate |
Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates |
|||
Ideal for storage and/or transportation of pH-sensitive materials, such as biological drug substance |
|
||
Key features | |||
Sterile and non-sterile options |
|||
Manufactured from a high-quality resins |
|||
Leakproof closures* |
|||
Triple-layer packaging (two heat seals and one carton liner) |
|
|
|
Produced in an ISO 7 clean room |
|
||
Compliant with USP <788> |
|
||
Certified to 1/3 of the allowable USP <788> particulate limits |
|
|
|
Washed with WFI (water for injection)** |
|
|
|
Inspected, assembled, and packaged in ISO 5 (Class 100) clean rooms with ISO 4 (Class 10) hoods and full gowning for operators |
|
|
* Our guarantee for a leakproof seal is subject to our standard product warranty, as set forth in the Thermo Fisher Scientific Terms and Conditions of Sale. Our products are leakproof under use with our required specifications at ambient temperature and pressure when used with their corresponding closures, when handled properly and secured by the user properly. However, to ensure safe usage, customers are advised to test our containers and closures under conditions of their planned application.
** Not all Platinum Certified Clean ultralow particulate bottles products require WFI wash to meet particulate claims
Fluid transfer systems play an important role in bioprocessing but can easily cause inefficiencies; multiple components need to be sourced, purchased, inventoried, and assembled. Our fit-for-purpose Thermo Scientific Nalgene Bottle and Carboy Assembly Systems are designed to reduce the burden of managing individual components and to enabled in-house system assembly and validation.
Available in a variety of resins and sizes
Closures and components to meet your needs
Supply assurance and risk mitigation
![]() |
Standard Single-Use Bottle Assembly Systems
|
![]() |
Customized Single-Use Bottle Assembly Systems
Contact us to learn more about how a custom bottle assembly system can benefit your laboratory.
|
![]() |
Customized Single-Use Carboy Assembly Systems
Ask a specialist about how a custom carboy assembly can meet your application needs. |
*** No leaks reported since 2021 launch as of the publishing this web page May 2024
![]() |
|
![]() |
Total Organic Carbon (TOC) Glassware
|
![]() |
|
![]() |
|
We understand the critical importance of maintaining clean and contaminant-free containers and component parts in the biopharma and vaccine industries. Our cleaning service offers unmatched expertise and state-of-the-art facilities to ensure the highest standards of cleanliness and compliance.
Designed for easy installation and maintenance, our Nalgene tanks and tank liners are cost-effective solutions that provide long-lasting performance, helping you optimize your fluid storage processes and minimize operational risks.
Looking for reliable containment to protect your high-quality products in storage, transport, and sampling? Made from high-quality resins, Nalgene production bottles and carboys offer leakproof* storage for sensitive liquids, buffers, media, and more. Nalgene containers help ensure your substances stay secure, with low and ultralow particulate options available, addressing particulate level (USP <788>), endotoxin level, and sterility requirements.
Enhance your bioprocessing knowledge with educational content to help you understand issues, solve problems, and make solid decisions.
Deep dive into a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
Learn, evaluate, connect.
Connect with a Bioprocessing representative or request more info.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.