Attune Flow Cytometers are compact benchtop cell-analyzers with hardware features that enable exceptionally fast sample flow rates, high clog resistance, and outstanding reliability. Attune instruments can be configured with up to 4 spatially-separated lasers and 14 fluorescence detectors. Acoustic focusing technology  enables sample flow rates up to 1,000 µL/min, ~10x times faster than traditional cytometers, without compromising data integrity. Attune Cytometric Software makes the system easy-to-use and offers an optional module for 21 CFR part 11 compliance.
The Attune CytPix model includes a high-speed brightfield camera to capture images of individual cells as they pass through the flow cell. Automated image analysis software produces morphometric data integrated with standard fluorescence parameters. These features enable researchers to enhance data precision and accuracy, and gain novel insights into their sample biology
Try our instrument configurator tool to design a system to fit your research needs.
New Invitrogen Attune Xenith Flow Cytometer
Fast, flexible, and spectrally brilliant—simplify high-speed, high-parameter analysis
Attune cell analyzer capabilities
Design your Attune system to fit your research goals
Try our instrument configurator tool to design a system built for your lab. 
Acoustic focusing cytometry
Sound waves enable precise cell alignment, resulting in exceptional data quality and ~10X faster sample flow rates.
Visualization of imaging-enhanced flow cytometry data measurement
Learn how the Edison-award winning Attune CytPix Flow Cytometer integrates imaging with flow cytometry to enhance data precision and accuracy.
Considerations when purchasing a flow cytometer
Learn about key experimental concepts and flow cytometer components.
Educational videos and webinars
Learn more from researchers who use Attune Flow Cytometers in their labs
FREE—Flow Cytometry Panel Design Service and Panel Builder Tool
Our scientists will design panels for you as many times you need—free of charge, no product purchase needed.
Workflow automation options
Pair your Attune flow cytometer with an autosampler for increased workflow efficiency, and add robotic integration for maximum throughput.