Biochemical ID Panels
Visibly faster biochemical ID with easy read panels
Get faster bacterial identification with ready prepared biochemical test panels. Perform multiple tests simultaneously for confidence in results. Visible color reactions and comparison to reactivity patterns stored in an extensive database aid interpretation and reduce subjectivity.
Browse Biochemical ID Panels
RapID Systems: better reporting, more coverage, advanced updates
Manual microbial identification is now easier than ever. With Thermo Scientific™ RapID™ Systems and ERIC™ Software, you’ll experience reduced turnaround time, visible color reactions and precise reporting for more organisms on more platforms.
Accurately identify more than 400 medically important organisms in less than four hours. RapID uses enzyme technology to reduce the time-to-results, with one-step inoculation and no oil or pipetting, providing a definitive answer to bacterial ID faster.
RapID systems are ideal for stand-alone testing or for complementing automated systems when testing for anaerobes, yeasts, Neisseria-Haemophilus, Enterobacteriaceae, staphylococci, streptococci, corynebacteria, non-fermenters, and urinary tract isolates.
Microbact Biochemical Systems: consistent volume, fewer reagents, easy read
Thermo Scientific™ Oxoid™ Microbact™ Biochemical Systems can be applied in a number of different industries, with applications for identifying Gram-negative bacilli (GNB), Listeria and staphylococci.
ID Panel Reagents
Thermo Scientific replacement reagents, ready-to-use inoculation fluid and accessories.
View all ID Panel Reagents
ID Panel Software
ERIC Software for RapID Systems
Designed exclusively for use with RapID Systems, the latest version of ERIC Software features an expanded taxonomy for unparalleled coverage and confidence. 
Microbact Identification Software
Easily read and interpret results from Microbact GNB, Listeria and Staphylococcus kits with the Microbact Software Program.
SmartNote: Why manual microorganism identification is critical for all microbiology laboratories
SmartNote: Why manual microorganism identification is critical for all microbiology laboratories
Accurate and definitive microorganism identification is essential for correct disease diagnosis, treatment of infection and epidemiological analysis. Biochemical tests are among the most important methods for microbial identification, even when a microbiology laboratory is using an automated system. Learn more about the benefits of manual ID, and how it can complement automated systems with our new SmartNote.
How to set up the RapID System