Search Thermo Fisher Scientific
Thermo Scientific BioProcess Containers (BPCs) and transfer assemblies are ready-to-use single-use flexible container systems for critical liquid handling applications in the biopharmaceutical and Biomanufacturing operations.
The Thermo Scientific single-use BPC product portfolio includes stocked catalog BPCs as well as custom BPCs in a variety of sizes and configurations that are manufactured at multiple facilities. This provides users with BPCs for at any step in the workflow and at any scale. Our multiple manufacturing sites help ensure an assurance of supply.
The benefits of ready-to-use single-use systems are well documented and include:
Films that are engineered to meet the most demanding requirements of your bioproduction processes.
Catalog 2D and 3D BPC products useful for a full range of applications are available with a variety of connection systems in sizes from 50 mL to 5,000 L.
Custom-configured BPCs to meet specific needs and specific processes including: storage, sampling, harvesting and separation; cell culture, powder containment and transportation applications.
Bring your bioproduction processes together seamlessly with an extensive range of catalog and customized fluid transfer products.
Support operations | Applications | BPC solution |
---|---|---|
Media or buffer preparation |
|
|
Filtration |
|
|
Mixing |
|
|
Harvest |
|
|
Filling |
|
|
Purification |
|
|
Bulk storage |
|
|
Waste collection |
|
|
Sampling |
|
|
Shipping |
|
|
Support operations | Applications | BPC solution |
---|---|---|
Media or buffer preparation |
|
|
Filtration |
|
|
Mixing |
|
|
Harvest |
|
|
Filling |
|
|
Purification |
|
|
Bulk storage |
|
|
Waste collection |
|
|
Sampling |
|
|
Shipping |
|
|
The Harvestainer BPC is part of a closed, self-contained, single-use solution for harvesting and separating cell cultures grown on microcarrier beads.
The Powdertainer II BPC is specifically designed for powder containment and dust-free powder transfer operations.
The Three60 Sampling System allows you to take samples of your product with minimal effort in a closed system.
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: