Search Thermo Fisher Scientific
Targeted next-generation sequencing (NGS) is a rapid and accurate solution for viral genomes, suitable for viral typing, epidemiological studies, and disease surveillance using archived samples from viral investigations.
Our viral typing NGS solutions provide ultra-high sensitivity, scalability, and rapid turnaround time, which are all vital to viral monitoring and investigation research.
Ion Torrent NGS technology is based on ultrahigh-multiplex PCR, the same technology that has enabled public health agencies to closely monitor viral infections.
The Ion AmpliSeq SARS-CoV-2 Insight Research Assay leverages a unique targeted sequencing approach that yields improved coverage and uniformity from samples previously unobtainable by NGS. Incorporation of intelligent panel design provides an increased tolerance to new mutations introduced within the viral genome. Integrated analysis tools enable automated consensus sequence generation, mutation annotation, and lineage assignment.
With Ion Torrent targeted NGS, easily go from DNA to data in less than 24 hours with ~45 minutes of hands-on time.Ion AmpliSeq libraries are prepared manually or automated with the Ion Chef System. Libraries are then placed in the Ion Chef System for emulsion PCR, enrichment, and loading onto Ion S5 Chips.
Select from pre-designed panels or configure your own with the Ion AmpliSeq Designer.
Select from pre-designed panels or configure your own with the Ion AmpliSeq Designer.
A major challenge for microbiologists and virologists is the prediction of patterns of evolution and emergence of disease agents. RNA viruses share the biological feature of high genetic variability, which causes them to appear as clouds of mutants. Viral variants also emerge through antigenic shift within animal reservoirs like bats, snakes, pigs, and birds.
Our Ion AmpliSeq panels are based on ultrahigh-multiplex PCR, the same technology that has enabled public health agencies to closely monitor viral infections.
A few of our NGS research panels for infectious disease research are highlighted below:
1. Grubaugh ND, et al. Genomic epidemiology reveals multiple introductions of Zika virus into the United States. Nature. 2017 Jun 15;546(7658):401-405.
2. Cullen M, et al. Deep sequencing of HPV16 genomes: A new high-throughput tool for exploring the carcinogenicity and natural history of HPV16 infection. Papillomavirus Res. 2015 Dec 1;1:3-11.
For Research Use Only. Not for use in diagnostic procedures.