Search Thermo Fisher Scientific
The Luminex family of systems encompasses four instrument models, each offering distinct multiplexing capabilities, throughput, and read times. These Luminex instruments operate on the quantitative fluorescent flow cytometry principle, leveraging the xMAP magnetic bead technology to detect small quantities of analytes. Used in combination with our ProcartaPlex multiplex assays for protein quantitation or QuantiGene Plex assays for gene expression analysis, the Luminex platform delivers a rapid and cost-effective immunoassay profiling tool. This synergy between Luminex instruments and our advanced multiplex assays enhances efficiency and precision in the detection and quantification of biomolecules.
xMAP INTELLIFLEX DR-SE System |
||||
Double the data with the second reporter channel | Compact footprint with our broadest dynamic range | Fast read-time and high-throughput | Fast and robust platform for up to 100 analytes | |
Performance | ||||
Multiplex capacity | Up to 500 Up to 2 parameters per analytes |
Up to 500 | Up to 500 | Up to 100 (80 on MagPlex) |
Dynamic range |
≥ 5.5 logs (RP1) ≥ 4.5 logs (RP2) |
≥ 5.5 logs (RP1) |
≥ 4.5 logs (RP1) |
≥ 3.5 logs (RP1) |
Read time |
96-well in ~20 min 384-well in ~75 min |
96-well in ~20 min 384-well in ~75 min |
96-well in ~20 min 384-well in ~75 min |
45 min (96-well) |
Sensitivity (assay dependent) |
0.06–1 pg/mL (Protein assays); 1,000 RNA copies per target/well (RNA assays) |
|||
Technical |
||||
Footprint including PC (linear bench space) |
58.4 cm x 61 cm x 76 cm |
58.4 cm x 61 cm x 76 cm |
110 cm x 62 cm x 63 cm |
115 cm x 60 cm x 50 cm |
Weight |
54.5 kg (120 lb) |
54.5 kg (120 lb) |
77.1 kg (170 lb) |
49 kg (113 lb) |
Optics |
Flow cytometry-based lasers, APDs, PMTs |
|||
Reporter laser/optics |
532 nm (green) and 405 nm (violet) | 532 nm (green) |
532 nm (green) |
532 nm (green) |
Automation-compatible hardware |
Front and side eject |
Front eject only |
Front eject only |
Front eject only |
Operational validation |
||||
Application |
||||
Luminex assays |
Protein assays/nucleic acid assays (ProcartaPlex / QuantiGene Plex) | |||
Sample volume |
6.3–80 µL (assay dependent) |
25–80 µL (assay dependent) |
||
Largest available panel |
80 |
|||
Plate format |
96-well or 384-well |
96-well or 384-well |
96-well or 384-well |
96-well |
Software |
||||
21 CFR compliance |
Yes | |||
Automation Module |
Yes |
Yes |
Integration software |
Integration software |
Analysis software |
Data analysis apps for Luminex assays on the Thermo Fisher Connect Platform |
xPONENT basic plus Thermo Fisher Connect Platform |
For Research Use Only. Not for use in diagnostic procedures.