Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001270491.1 | NP_001257420.1 | ||||
NM_012423.3 | NP_036555.1 | |||||
NR_073024.1 | ||||||
GenBank mRNA | AB082924.1 | - | ||||
AK056837.1 | - | |||||
AK130605.1 | - | |||||
AK222747.1 | - | |||||
AK291120.1 | - | |||||
AK297859.1 | - | |||||
BC000514.2 | - | 2822802 | ||||
BC000847.1 | - | |||||
BC001675.2 | - | 2961667 | ||||
BC001836.2 | - | 2961667 | ||||
BC004900.2 | - | |||||
BC013230.1 | - | |||||
BC032107.2 | - | |||||
BC062537.1 | - | 4908022 | ||||
BC065236.1 | - | 6161599 | ||||
BC070223.1 | - | 6736442 | ||||
BC071929.1 | - | 5808487 | ||||
BC105605.1 | - | 6726477 | ||||
BG536330.1 | - | |||||
BQ069994.1 | - | |||||
ENST00000391857.0 | - | |||||
X56932.1 | - |
Set Membership: |
![]() |