Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTB EBGEX Assays ›
Gene Symbol:
Actb
Gene Name:
actin, beta
Gene Aliases:
Actx, E430023M04Rik, beta-actin
Chromosome Location:
Chr.5: 142903116 - 142906724 on Build GRCm38
UniGene ID:
Mm.328431
Species:
Mouse
Species Specific ID (Flybase ID):
-
Type:
4
Detection Label:
Alexa Fluor 488
ProductType:
PrimeFlow Probe Set
Assay ID
VB4-10432-PF
Size
40 tests
100 tests
Availability
Made To Order
Catalog #
PF-204
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Actb
Interrogated Sequence
Translated Protein
IMAGE Clone ID
GenBank mRNA
AB028847.1
-
AK075973.1
-
AK078935.1
-
AK088287.1
-
AK088691.1
-
AK145191.1
-
AK145196.1
-
AK145308.1
-
AK147787.1
-
AK149920.1
-
AK150545.1
-
AK150553.1
-
AK150558.1
-
AK150570.1
-
AK150662.1
-
AK150675.1
-
AK150711.1
-
AK150854.1
-
AK150866.1
-
AK150879.1
-
AK151010.1
-
AK151136.1
-
AK151145.1
-
AK151159.1
-
AK151166.1
-
AK151190.1
-
AK151202.1
-
AK151226.1
-
AK151277.1
-
AK151297.1
-
AK151350.1
-
AK151385.1
-
AK151386.1
-
AK151398.1
-
AK151444.1
-
AK151468.1
-
AK151514.1
-
AK151663.1
-
AK151712.1
-
AK151898.1
-
AK151956.1
-
AK151995.1
-
AK151999.1
-
AK152043.1
-
AK152052.1
-
AK152083.1
-
AK152218.1
-
AK152433.1
-
AK152470.1
-
AK152591.1
-
AK152615.1
-
AK152651.1
-
AK152671.1
-
AK152813.1
-
AK152841.1
-
AK152844.1
-
AK153461.1
-
AK159759.1
-
AK159834.1
-
AK160029.1
-
AK166349.1
-
AK166498.1
-
AK167117.1
-
AK167825.1
-
AK167960.1
-
AK177309.1
-
AK179836.1
-
AK180583.1
-
AK180740.1
-
AK180898.1
-
AK184086.1
-
AK184228.1
-
AK184732.1
-
AK187650.1
-
AK187727.1
-
AK189351.1
-
AK196898.1
-
AK197508.1
-
AK198515.1
-
AK199846.1
-
AK200180.1
-
AK200883.1
-
AK201181.1
-
AK202772.1
-
AK203791.1
-
AK204356.1
-
AK206771.1
-
AK208252.1
-
AK208556.1
-
AK209954.1
-
AK212447.1
-
AK212873.1
-
AK215935.1
-
AK216753.1
-
AK218610.1
-
AK218630.1
-
AK220069.1
-
AY618569.1
-
BC040513.1
-
BC138611.1
-
8861631
BC138614.1
-
8861634
BE992124.1
-
BY783897.1
-
CX211008.1
-
EF095208.1
-
J04181.1
-
M12481.1
-
U89400.1
-
X03672.1
-
X03765.1
-
Back To Top
More Information
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-5xwc9:80/10.239.165.207:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0