Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023733.hg.1 |
TC03001374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023733.hg.1 |
ENST00000302506.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023733.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000351231.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000437972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
ENST00000459900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011009.hg.1 |
NM_201567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
ENST00000302506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
ENST00000443342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
NM_001789.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300180501.hg.1 |
TC0300011009.hg.1 |
Clariom D TC |