Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002244.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002244.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002244.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002251.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002251.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002251.hg.1 |
ENST00000535674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002251.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002251.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002252.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002252.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002252.hg.1 |
ENST00000535674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002252.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002252.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002246.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002246.hg.1 |
ENST00000535674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002246.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002246.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002250.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002250.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002250.hg.1 |
ENST00000535674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002250.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002250.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002248.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002248.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002248.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002248.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002247.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002247.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002247.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002249.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002249.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002249.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002249.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002253.hg.1 |
ENST00000228872.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002253.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002253.hg.1 |
ENST00000535674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002253.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002253.hg.1 |
TC12000178.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200148120.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148120.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148120.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148120.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148120.hg.1 |
TC1200006888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200148118.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148118.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148118.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148118.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148118.hg.1 |
TC1200006888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148125.hg.1 |
TC1200006888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200148119.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148119.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148119.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148119.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148119.hg.1 |
TC1200006888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148124.hg.1 |
TC1200006888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
ENST00000477087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006888.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
ENST00000228872.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
ENST00000396340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
ENST00000442489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
NM_004064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148123.hg.1 |
TC1200006888.hg.1 |
Clariom D TC |