Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004821.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004821.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004821.hg.1 |
TC22000259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004818.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004818.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004818.hg.1 |
TC22000259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004820.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004820.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004820.hg.1 |
TC22000259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004816.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004816.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004816.hg.1 |
TC22000259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004822.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004822.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004822.hg.1 |
TC22000259.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150627.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150624.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150626.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150635.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150636.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
ENST00000494998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007204.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150632.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
TC2200007204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
NM_002133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
ENST00000216117.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
ENST00000412893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
ENST00000481190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150634.hg.1 |
NM_002133.2 |
Refseq |