Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000000412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000536844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000537621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000537621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000537621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000537621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000537621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000537621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000539143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000539143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000539143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000539143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000539143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000540837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000540837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000540837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000540837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000540837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000540837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000541507.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000543845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
ENST00000544245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_001207024.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009843.hg.1 |
NM_002355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008122.hg.1 |
ENST00000436086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008122.hg.1 |
ENST00000436086.2 |
Ensembl |