Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000596593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000596593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009369.hg.1 |
ENST00000317082.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009369.hg.1 |
ENST00000404338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009369.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009369.hg.1 |
TC19000669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009371.hg.1 |
ENST00000404338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009371.hg.1 |
ENST00000501576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009371.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009371.hg.1 |
TC19000669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009370.hg.1 |
ENST00000404338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009370.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009370.hg.1 |
TC19000669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009368.hg.1 |
ENST00000317082.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009368.hg.1 |
ENST00000404338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009368.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009368.hg.1 |
TC19000669.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
TC1900008399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164526.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
TC1900008399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164527.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000596593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000596593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008399.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
TC1900008399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000598548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164523.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
TC1900008399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000599284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164525.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
NM_004491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
TC1900008399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000404338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000595822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000614079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
ENST00000615647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164528.hg.1 |
NM_004491.4 |
Refseq |