Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001524.hg.1 |
TC08000149.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001524.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001524.hg.1 |
ENST00000535763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001524.hg.1 |
ENST00000538071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001524.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001526.hg.1 |
TC08000149.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001526.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001526.hg.1 |
ENST00000535763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001526.hg.1 |
ENST00000538071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001526.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001525.hg.1 |
TC08000149.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001525.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001525.hg.1 |
ENST00000535763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001525.hg.1 |
ENST00000538071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001525.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146957.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146957.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146957.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146957.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146957.hg.1 |
TC0800006927.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000519773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000520959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000521994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000522701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000523696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
ENST00000524029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006927.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146958.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146958.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146958.hg.1 |
ENST00000311322.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146958.hg.1 |
NM_000237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146958.hg.1 |
TC0800006927.hg.1 |
Clariom D TC |