Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025542.hg.1 |
TC06002043.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025542.hg.1 |
ENST00000368626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025542.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025540.hg.1 |
TC06002043.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025540.hg.1 |
ENST00000368626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025540.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025541.hg.1 |
TC06002043.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025541.hg.1 |
ENST00000368626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025541.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189424.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189424.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189424.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189424.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189424.hg.1 |
TC0600012952.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189427.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189427.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189427.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189427.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189427.hg.1 |
TC0600012952.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189426.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189426.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189426.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189426.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189426.hg.1 |
TC0600012952.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012952.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189425.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189425.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189425.hg.1 |
ENST00000606080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189425.hg.1 |
NM_002031.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189425.hg.1 |
TC0600012952.hg.1 |
Clariom D TC |