Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025953.hg.1 |
ENST00000301634.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025953.hg.1 |
ENST00000405273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025953.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025953.hg.1 |
TC17001914.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700196985.hg.1 |
TC1700011892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000301634.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000586613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000588734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000590862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
ENST00000592126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011892.hg.1 |
NM_003258.4 |
Refseq |