Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180015.hg.1 |
TC0600011661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |