Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013514.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013514.hg.1 |
ENST00000503833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013514.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013514.hg.1 |
TC20000992.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000472025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009646.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
NM_001336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
TC2000009646.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
ENST00000217131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
ENST00000488395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
ENST00000503833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167293.hg.1 |
NM_001336.3 |
Refseq |