Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024186.hg.1 |
ENST00000239138.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024186.hg.1 |
ENST00000439174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024186.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024186.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024186.hg.1 |
TC17001808.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011476.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
ENST00000439174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
NM_006572.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192907.hg.1 |
TC1700011476.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700192910.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192910.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192910.hg.1 |
ENST00000541118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192910.hg.1 |
NM_001282425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192910.hg.1 |
TC1700011476.hg.1 |
Clariom D TC |