Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000357266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000452048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000452048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000452048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000452048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000536438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000539068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
ENST00000542713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145776.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_001145777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NM_004117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011635.hg.1 |
NR_027117.2 |
Refseq |